Assay Details
| Gene Symbol: Gpaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010331.2 | NP_034461.1 | 4 - 5 | 606 | 95 | |
| GenBank mRNA | AB002136.1 | - | 4 - 5 | 535 | 95 | |
| AB006970.1 | - | 4 - 5 | 542 | 95 | ||
| AK077518.1 | - | 4 - 5 | 606 | 95 | ||
| AK164219.1 | - | 4 - 5 | 552 | 95 | ||
| AK164978.1 | - | 4 - 5 | 606 | 95 | ||
| AK168057.1 | - | 4 - 5 | 809 | 95 | ||
| AK192435.1 | - | 1 - 2 | 103 | 95 | ||
| BC006697.1 | - | 4 - 5 | 545 | 3498573 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347594 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347600 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347597 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347596 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347601 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166779 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171190 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30347614 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171190 | - | - |
| Whole Mouse Genome G4122A | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | BC006697.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AK164219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AK164978.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AK168057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273203 | AB006970.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107530 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122301 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12289185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12289191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12289186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12289192 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347594 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347596 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347600 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347597 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30347601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30347604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12078036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12078069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12078064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30347604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424811 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424822 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455954_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448225_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424810 | - | - |
| Mouse Genome 430 2.0 Array | 1448225_at | - | - |
| Mouse Genome 430 2.0 Array | 1455954_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438152_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448225_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441004_at | - | - |
| Mouse Genome 430 2.0 Array | 1455954_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1441004_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438152_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14731_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14731_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500004055.mm.1 | NM_010331.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004055.mm.1 | NM_010331.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004055.mm.1 | TC1500000585.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455954_x_at | - | - |
| Mouse Expression Set 430 | 1448225_at | - | - |
| Mouse Expression Set 430 | 1455954_PM_x_at | - | - |
| Mouse Expression Set 430 | 1448225_PM_at | - | - |
| Murine Genome U74v2 Set | 161682_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100911_PM_at | - | - |
| Murine Genome U74v2 Set | 161682_f_at | - | - |
| Murine Genome U74v2 Set | 100911_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448225_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455954_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448225_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455954_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455954_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455954_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448225_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448225_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 931846 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1500463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500463 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255423 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255423 | - | - |
| mouse-6 v1.1 (Illumina) | scl47800.9.47_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500463 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47800.9.47_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500463 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255423 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255423 | - | - |

Gpaa1
Genomic Map