Assay Details
| Gene Symbol: Gpaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010331.2 | NP_034461.1 | 7 - 8 | 1112 | 73 | |
| GenBank mRNA | AB002136.1 | - | 7 - 8 | 1041 | 73 | |
| AB006970.1 | - | 7 - 8 | 1048 | 73 | ||
| AK077518.1 | - | 7 - 8 | 1112 | 73 | ||
| AK164219.1 | - | 7 - 8 | 1058 | 73 | ||
| AK164978.1 | - | 7 - 8 | 1112 | 73 | ||
| AK168057.1 | - | 7 - 8 | 1315 | 73 | ||
| BC006697.1 | - | 7 - 8 | 1051 | 3498573 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpaa1
Genomic Map