Assay Details
| Gene Symbol: Gpx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010343.2 | NP_034473.2 | 4 - 5 | 485 | 137 | |
| GenBank mRNA | AK029731.1 | - | 4 - 5 | 486 | 137 | |
| BC100749.1 | - | 4 - 5 | 485 | 40046457 | 137 | |
| BC103588.1 | - | 4 - 5 | 485 | 40045838 | 137 | |
| BC103589.1 | - | 4 - 5 | 485 | 40045837 | 137 | |
| BC105648.1 | - | 4 - 5 | 485 | 40046458 | 137 | |
| X53780.1 | - | 3 - 4 | 321 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166758 | X53780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166758 | NM_010343.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166758 | AK033841.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596450 | NM_007703.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596450 | U97107.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415416 | AK039339.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415416 | AK052852.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935388 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146871 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166758 | X53780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166758 | NM_010343.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166758 | AK033841.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267644 | X53780.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28872119 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146871 | - | - |
| Whole Mouse Genome G4122A | A_51_P267644 | X53780.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124225 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126753 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146871 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10511535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10774886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10774889 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10774901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28872090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11011362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11008158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10459815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10774901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10728195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10801478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10459820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10536918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10981841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11050739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10459813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10459821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28872098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408005 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420698_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407999 | - | - |
| Mouse Genome 430 2.0 Array | 1420698_at | - | - |
| Mouse Genome 430 2.0 Array | 1420698_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14780_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14780_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300009711.mm.1 | NM_010343.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009711.mm.1 | NM_010343.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009711.mm.1 | TC1300001534.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009712.mm.1 | NM_010343.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009712.mm.1 | NM_010343.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009712.mm.1 | TC1300001534.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420698_at | - | - |
| Mouse Expression Set 430 | 1420698_PM_at | - | - |
| Murine Genome U74v2 Set | 94692_PM_at | - | - |
| Murine Genome U74v2 Set | 94692_at | - | - |
| Murine Genome U74v2 Set | 161986_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161986_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420698_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420698_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420698_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420698_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 879640 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690739 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638087 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773355 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773355 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2638087 | - | - |
| mouse-6 v1.1 (Illumina) | scl44241.5_198-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003700.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200397 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44241.5_198-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003700.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690739 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2638087 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2902167 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2902162 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2902167 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2638087 | - | - |

Gpx5
Genomic Map