Assay Details
| Gene Symbol: Gsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010353.2 | NP_034483.1 | 1 | 153 | 62 | |
| GenBank mRNA | AK138750.1 | - | 1 | 154 | 62 | |
| BC113148.1 | - | 1 | 147 | 40054435 | 62 | |
| D87326.1 | - | 1 | 153 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cell cycle
mitotic sister chromatid cohesion
phosphorylation
chromatin modification
intracellular signal transduction
protein localization to chromosome, centromeric region
regulation of spindle checkpoint
negative regulation of G1/S transition of mitotic cell cycle
histone H3-T3 phosphorylation involved in chromosome passenger complex localization to kinetochore
nucleotide binding
DNA binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
histone kinase activity (H3-T3 specific)
protein phosphorylation
cell cycle
mitotic sister chromatid cohesion
phosphorylation
chromatin modification
intracellular signal transduction
protein localization to chromosome, centromeric region
regulation of spindle checkpoint
negative regulation of G1/S transition of mitotic cell cycle
histone H3-T3 phosphorylation involved in chromosome passenger complex localization to kinetochore
nucleotide binding
DNA binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
histone kinase activity (H3-T3 specific)
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915402 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915404 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915400 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P151586 | D87326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P151586 | D87326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124413 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P151586 | D87326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P151586 | D87326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P151586 | D87326.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P151586 | D87326.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27915422 | - | - |
| Whole Mouse Genome G4122A | A_51_P151586 | D87326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P151586 | D87326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P151586 | NM_010353.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121979 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09717655 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09717680 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915400 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915402 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27915404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27915406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09663357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09717648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09717682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09663359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09672304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09487183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09672307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09672302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27915432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09717670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388236 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388237 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450886_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388234 | - | - |
| Mouse Genome 430 2.0 Array | 1450886_at | - | - |
| Mouse Genome 430 2.0 Array | 1450886_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14841_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14841_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100029250.mm.1 | NM_010353.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029250.mm.1 | NM_010353.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029250.mm.1 | TC1100003156.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450886_PM_at | - | - |
| Mouse Expression Set 430 | 1450886_at | - | - |
| Murine Genome U74v2 Set | 99522_at | - | - |
| Murine Genome U74v2 Set | 99522_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450886_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450886_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450886_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450886_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 473479 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5270270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235363 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235363 | - | - |
| mouse-6 v1.1 (Illumina) | scl39961.1.1_178-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39961.1.1_178-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1235363 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235363 | - | - |

Gsg2
Genomic Map