Assay Details
| Gene Symbol: Gsta4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010357.3 | NP_034487.2 | 2 - 3 | 201 | 129 | |
| GenBank mRNA | AK008189.1 | - | 2 - 3 | 187 | 129 | |
| AK008193.1 | - | 3 - 4 | 152 | 129 | ||
| AK008400.1 | - | 2 - 3 | 185 | 129 | ||
| AK008490.1 | - | 2 - 3 | 152 | 129 | ||
| AK009668.1 | - | 2 - 3 | 155 | 129 | ||
| AK010098.1 | - | 2 - 3 | 155 | 129 | ||
| AK011177.1 | - | 2 - 3 | 152 | 129 | ||
| AK011841.1 | - | 2 - 3 | 180 | 129 | ||
| AK019271.1 | - | 2 - 3 | 176 | 129 | ||
| BC012639.1 | - | 2 - 3 | 155 | 3995378 | 129 | |
| BU953110.1 | - | 2 - 3 | 204 | 129 | ||
| L06047.1 | - | 2 - 3 | 153 | 129 | ||
More Information
Set Membership: |
|

Gsta4
Genomic Map