Assay Details
| Gene Symbol: Gtf2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080746.2 | NP_001074215.1 | 23 - 24 | 2453 | 66 | |
| NM_001080747.2 | NP_001074216.1 | 22 - 23 | 2390 | 66 | ||
| NM_001080748.2 | NP_001074217.1 | 21 - 22 | 2333 | 66 | ||
| NM_001080749.2 | NP_001074218.1 | 21 - 22 | 2333 | 66 | ||
| NM_010365.4 | NP_034495.2 | 22 - 23 | 2396 | 66 | ||
| GenBank mRNA | AF017085.1 | - | 23 - 24 | 2097 | 66 | |
| AK145698.1 | - | 13 - 14 | 1281 | 66 | ||
| AK147201.1 | - | 21 - 22 | 2291 | 66 | ||
| AY030290.1 | - | 22 - 23 | 2083 | 66 | ||
| AY030291.1 | - | 22 - 23 | 2089 | 66 | ||
| AY030292.1 | - | 21 - 22 | 2026 | 66 | ||
| AY030293.1 | - | 23 - 24 | 2146 | 66 | ||
| BC047387.1 | - | 21 - 22 | 1981 | 66 | ||
| BC053044.1 | - | 23 - 24 | 2422 | 5704935 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2i
Genomic Map