Assay Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252266.1 | NP_001239195.1 | 5 - 6 | 653 | 95 | |
| NM_010492.3 | NP_034622.3 | 6 - 7 | 1066 | 95 | ||
| GenBank mRNA | AB303567.1 | - | 5 - 6 | 577 | 95 | |
| AK016989.1 | - | 6 - 7 | 769 | 95 | ||
| BC002030.1 | - | 6 - 7 | 700 | 3487814 | 95 | |
| CB056959.1 | - | 1 - 2 | 163 | 95 | ||
| DV659030.1 | - | 4 - 5 | 438 | 95 | ||
| U26459.1 | - | 6 - 7 | 689 | 95 | ||
| U26460.1 | - | 5 - 6 | 595 | 95 | ||
| U26461.1 | - | 5 - 6 | 594 | 95 | ||
| U34595.1 | - | 4 - 5 | 562 | 95 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BP774450.1 | - | Chr. - - [-] on Build | 95 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ica1
Genomic Map