Assay Details
| Gene Symbol: Icam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010494.1 | NP_034624.1 | 2 - 3 | 484 | 82 | |
| GenBank mRNA | AK007801.1 | - | 2 - 3 | 419 | 82 | |
| BC039128.1 | - | 2 - 3 | 416 | 3962648 | 82 | |
| BC039970.1 | - | 2 - 3 | 436 | 4487248 | 82 | |
| X65493.1 | - | 2 - 3 | 484 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982049 | BC039970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982049 | X65493.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982049 | NM_010494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982049 | BC039128.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133794 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125206 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982049 | BC039970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982049 | X65493.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982049 | NM_010494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982049 | BC039128.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267754 | BC039970.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267754 | X65493.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267754 | NM_010494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267754 | AK007801.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267754 | BC039128.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28101305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125206 | - | - |
| Whole Mouse Genome G4122A | A_51_P267754 | BC039970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267754 | X65493.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267754 | NM_010494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267754 | AK007801.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267754 | BC039128.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138593 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125206 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09146074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09146071 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09146065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09146063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09146058 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09146068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28101324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08994376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09127361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09199698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08994366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09499612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08994378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09146058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09199695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09056820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09540966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09339003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09146074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09691416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09146068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09499604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09146072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09146062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09460692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09445211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09542277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28101265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10392178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392181 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448862_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10392177 | - | - |
| Mouse Genome 430 2.0 Array | 1448862_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448862_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15896_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15896_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100036759.mm.1 | NM_010494.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100036759.mm.1 | TC1100003923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100036758.mm.1 | NM_010494.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100036758.mm.1 | TC1100003923.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448862_PM_at | - | - |
| Mouse Expression Set 430 | 1448862_at | - | - |
| Murine Genome U74v2 Set | 99053_PM_at | - | - |
| Murine Genome U74v2 Set | 99053_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448862_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448862_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448862_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448862_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6400113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6400113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618408 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618408 | - | - |
| mouse-6 v1.1 (Illumina) | scl39447.7.1_219-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39447.7.1_219-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2618408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618408 | - | - |

Icam2
Genomic Map