Assay Details
| Gene Symbol: Ids | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010498.3 | NP_034628.2 | 2 - 3 | 426 | 73 | |
| GenBank mRNA | AF538870.1 | - | 2 - 3 | 425 | 73 | |
| AK085237.1 | - | 3 - 4 | 303 | 73 | ||
| AK132287.1 | - | 3 - 4 | 73 | |||
| AK166178.1 | - | 2 - 3 | 304 | 73 | ||
| AK166443.1 | - | 3 - 4 | 255 | 73 | ||
| L07921.1 | - | 2 - 3 | 303 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ids
Genomic Map