Assay Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010581.3 | NP_034711.1 | 2 - 3 | 532 | 88 | |
| GenBank mRNA | AB012693.1 | - | 2 - 3 | 471 | 88 | |
| BC012667.1 | - | 2 - 3 | 466 | 4187965 | 88 | |
| Z25524.1 | - | 2 - 3 | 532 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd47
Genomic Map