Assay Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010630.2 | NP_034760.2 | 17 - 18 | 2151 | 78 | |
| GenBank mRNA | AK144304.1 | - | 18 - 19 | 2310 | 78 | |
| D49545.1 | - | 17 - 18 | 2227 | 78 | ||
| U92949.1 | - | 17 - 18 | 2141 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349812 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349811 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349814 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349813 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349764 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149500 | NM_010630.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12528 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178127 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149500 | NM_010630.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P505689 | U92949.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349781 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03074356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178127 | - | - |
| Whole Mouse Genome G4122A | A_51_P505689 | U92949.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132701 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113355 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349764 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349811 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349813 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349814 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349812 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11924435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12230060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12089218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12181080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11924424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12230067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349784 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424964 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421312_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426493_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424945 | - | - |
| Mouse Genome 430 2.0 Array | 1426493_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426493_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421312_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421312_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16581_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16581_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500004363.mm.1 | NM_010630.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004363.mm.1 | NM_010630.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004363.mm.1 | TC1500000603.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500004362.mm.1 | NM_010630.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004362.mm.1 | NM_010630.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004362.mm.1 | TC1500000603.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421312_a_at | - | - |
| Mouse Expression Set 430 | 1421312_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426493_a_at | - | - |
| Mouse Expression Set 430 | 1426493_PM_a_at | - | - |
| Murine Genome U74v2 Set | 93891_PM_at | - | - |
| Murine Genome U74v2 Set | 93890_PM_at | - | - |
| Murine Genome U74v2 Set | 93890_at | - | - |
| Murine Genome U74v2 Set | 93891_at | - | - |
| Murine Genome U74v2 Set | 170120_r_at | - | - |
| Murine Genome U74v2 Set | 170120_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426493_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421312_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421312_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426493_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421312_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421312_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426493_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426493_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 532222 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3450056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003450056 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2695158 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2695158 | - | - |
| mouse-6 v1.1 (Illumina) | scl016581.18_49-S | - | - |
| mouse-6 v1.1 (Illumina) | scl47789.18.22_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170500 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3450056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47789.18.22_35-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016581.18_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450056 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2695158 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613504 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2695158 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613504 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl016581.18_49-S | - | - |

Kifc2
Genomic Map