Assay Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010654.3 | NP_034784.1 | 3 - 4 | 185 | 62 | |
| GenBank mRNA | AF030311.1 | - | 2 - 3 | 165 | 62 | |
| AF030312.1 | - | 3 - 4 | 214 | 62 | ||
| AF039025.1 | - | 3 - 4 | 179 | 62 | ||
| AF057714.1 | - | 2 - 3 | 165 | 62 | ||
| AK136548.1 | - | 3 - 4 | 186 | 62 | ||
| AK162976.1 | - | 3 - 4 | 187 | 62 | ||
| BC117112.1 | - | 3 - 4 | 203 | 40126431 | 62 | |
| BC120854.1 | - | 3 - 4 | 205 | 40129777 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113498 | AF057714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113498 | AK136548.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113498 | AK162976.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113498 | NM_010654.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113498 | AF030311.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | AK132906.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | AK140808.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | AK144737.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | AK179734.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | BC013898.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | NM_009878.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | U19597.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | U20497.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827619 | XM_001481149.1 | refseq_mrna_mouse_XM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107524 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177114 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113498 | AF057714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113498 | AK136548.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113498 | AK162976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113498 | NM_010654.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113498 | AF030311.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280947 | AF057714.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280947 | AK136548.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280947 | AK162976.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280947 | BC117112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P280947 | BC120854.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24844433 | - | - |
| Whole Mouse Genome G4122A | A_51_P280947 | AF057714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P280947 | AK136548.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P280947 | AK162976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P280947 | BC117112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P280947 | BC120854.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05275100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05275102 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05275117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05430722 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05522722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24844433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05557627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05844895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05557630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05726214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05290098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05806669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05290097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05109613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24844448 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542217 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542219 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542216 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542218 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542215 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460245_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542214 | - | - |
| Mouse Genome 430 2.0 Array | 1460245_at | - | - |
| Mouse Genome 430 2.0 Array | 1460245_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16643_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16643_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012547.mm.1 | TC0600001538.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012547.mm.1 | NM_010654.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012547.mm.1 | NM_010654.3 | Refseq |
| Mouse Expression Set 430 | 1460245_at | - | - |
| Mouse Expression Set 430 | 1460245_PM_at | - | - |
| Murine Genome U74v2 Set | 93677_PM_at | - | - |
| Murine Genome U74v2 Set | 93677_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460245_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460245_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460245_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460245_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 597390 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3190114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190114 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255860 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255860 | - | - |
| mouse-6 v1.1 (Illumina) | scl29460.6.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29460.6.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255860 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255860 | - | - |

Klrd1
Genomic Map