Assay Details
| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010657.3 | NP_034787.2 | 4 - 5 | 5880 | 89 | |
| GenBank mRNA | AK173200.1 | - | 1 - 2 | 1126 | 89 | |
| AY454345.1 | - | 5 - 6 | 89 | |||
| BC145272.1 | - | 1 - 2 | 5399 | 89 | ||
| L07911.1 | - | 1 - 2 | 89 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Hivep3
Genomic Map