Assay Details
| Gene Symbol: Ldha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136069.2 | NP_001129541.2 | 1 - 2 | 314 | 72 | |
| GenBank mRNA | BY195537.1 | - | 1 - 2 | 316 | 72 | |
| U13687.1 | - | 1 - 2 | 181 | 72 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the U13687.1 transcripts.
Set Membership: |
|

Ldha
Genomic Map