Assay Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010718.4 | NP_034848.1 | 3 - 4 | 433 | 69 | |
| NM_173053.1 | NP_774958.1 | 2 - 3 | 564 | 69 | ||
| GenBank mRNA | AB005131.1 | - | 3 - 4 | 286 | 69 | |
| AB005132.1 | - | 2 - 3 | 548 | 69 | ||
| AB008117.1 | - | 3 - 4 | 369 | 69 | ||
| AK146099.1 | - | 2 - 3 | 522 | 69 | ||
| BC007129.1 | - | 3 - 4 | 376 | 3496836 | 69 | |
| BC086629.1 | - | 2 - 3 | 496 | 69 | ||
| CV784400.1 | - | 2 - 3 | 564 | 69 | ||
| U88618.1 | - | 3 - 4 | 286 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Limk2
Genomic Map