Assay Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010718.4 | NP_034848.1 | 4 - 5 | 538 | 53 | |
| NM_173053.1 | NP_774958.1 | 3 - 4 | 669 | 53 | ||
| GenBank mRNA | AB005131.1 | - | 4 - 5 | 391 | 53 | |
| AB005132.1 | - | 3 - 4 | 653 | 53 | ||
| AB008117.1 | - | 4 - 5 | 474 | 53 | ||
| AK146099.1 | - | 3 - 4 | 627 | 53 | ||
| AK188227.1 | - | 2 - 3 | 243 | 53 | ||
| BC007129.1 | - | 4 - 5 | 481 | 3496836 | 53 | |
| BC086629.1 | - | 3 - 4 | 601 | 53 | ||
| U88618.1 | - | 4 - 5 | 391 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Limk2
Genomic Map