Assay Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010718.4 | NP_034848.1 | 5 - 6 | 728 | 62 | |
| NM_173053.1 | NP_774958.1 | 4 - 5 | 859 | 62 | ||
| GenBank mRNA | AB005131.1 | - | 5 - 6 | 581 | 62 | |
| AB005132.1 | - | 4 - 5 | 843 | 62 | ||
| AB008117.1 | - | 5 - 6 | 664 | 62 | ||
| AK146099.1 | - | 4 - 5 | 817 | 62 | ||
| AK182337.1 | - | 1 - 2 | 63 | 62 | ||
| BC007129.1 | - | 5 - 6 | 671 | 3496836 | 62 | |
| BC086629.1 | - | 4 - 5 | 791 | 62 | ||
| U88618.1 | - | 5 - 6 | 581 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Limk2
Genomic Map