Assay Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034030.2 | NP_001029202.1 | 6 - 7 | 800 | 70 | |
| NM_010718.4 | NP_034848.1 | 10 - 11 | 1439 | 70 | ||
| NM_173053.1 | NP_774958.1 | 9 - 10 | 1570 | 70 | ||
| GenBank mRNA | AB005131.1 | - | 10 - 11 | 1292 | 70 | |
| AB005132.1 | - | 9 - 10 | 1554 | 70 | ||
| AB008117.1 | - | 10 - 11 | 1375 | 70 | ||
| AB012092.1 | - | 6 - 7 | 800 | 70 | ||
| AB012291.1 | - | 6 - 7 | 800 | 70 | ||
| AK034900.1 | - | 1 - 2 | 101 | 70 | ||
| AK146099.1 | - | 9 - 10 | 1527 | 70 | ||
| BC007129.1 | - | 10 - 11 | 1382 | 3496836 | 70 | |
| BC086629.1 | - | 9 - 10 | 1500 | 70 | ||
| U88618.1 | - | 10 - 11 | 1292 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Limk2
Genomic Map