Assay Details
| Gene Symbol: Muc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010739.2 | NP_034869.1 | 1 - 2 | 97 | 99 | |
| GenBank mRNA | J04634.1 | - | 1 - 2 | 97 | 99 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the J04634.1,NM_010739.2 transcripts.
Set Membership: |
|

Muc13
Genomic Map