Assay Details
| Gene Symbol: Muc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010739.2 | NP_034869.1 | 4 - 5 | 1012 | 107 | |
| GenBank mRNA | AK132274.1 | - | 3 - 4 | 857 | 107 | |
| BC024321.1 | - | 3 - 4 | 978 | 4219376 | 107 | |
| J04634.1 | - | 4 - 5 | 1012 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Muc13
Genomic Map