| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200039101.mm.1 |
NM_010762.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039101.mm.1 |
NM_010762.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039101.mm.1 |
TC0200004563.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039102.mm.1 |
NM_001171187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039102.mm.1 |
NM_001171187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039102.mm.1 |
NM_010762.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039102.mm.1 |
NM_010762.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039102.mm.1 |
TC0200004563.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039103.mm.1 |
NM_001171187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039103.mm.1 |
NM_001171187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039103.mm.1 |
NM_010762.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039103.mm.1 |
NM_010762.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039103.mm.1 |
TC0200004563.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039104.mm.1 |
NM_001171187.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039104.mm.1 |
NM_001171187.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039104.mm.1 |
NM_010762.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039104.mm.1 |
NM_010762.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039104.mm.1 |
TC0200004563.mm.1 |
MTA TC |