Assay Details
| Gene Symbol: Mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010762.5 | NP_034892.1 | 2 - 3 | 826 | 60 | |
| GenBank mRNA | AK004639.1 | - | 2 - 3 | 413 | 60 | |
| AK158653.1 | - | 2 - 3 | 827 | 60 | ||
| AK169172.1 | - | 2 - 3 | 350 | 60 | ||
| BC006826.1 | - | 2 - 3 | 323 | 3597561 | 60 | |
| Y07626.1 | - | 2 - 3 | 302 | 60 | ||
| Y07812.1 | - | 2 - 3 | 264 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mal
Genomic Map