Assay Details
| Gene Symbol: Osmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011019.3 | NP_035149.2 | 15 - 16 | 2329 | 71 | |
| GenBank mRNA | AB015978.1 | - | 15 - 16 | 2980 | 71 | |
| AF058805.1 | - | 15 - 16 | 2299 | 71 | ||
| AK157508.1 | - | 10 - 11 | 1502 | 71 | ||
| BC137703.1 | - | 14 - 15 | 2227 | 8860722 | 71 | |
More Information
Set Membership: |
|

Osmr
Genomic Map