Assay Details
| Gene Symbol: Slc23a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011397.4 | NP_035527.3 | 10 - 11 | 1288 | 71 | |
| GenBank mRNA | AF058318.1 | - | 10 - 11 | 1198 | 71 | |
| AK085499.1 | - | 10 - 11 | 1203 | 71 | ||
| AK143959.1 | - | 10 - 11 | 1289 | 71 | ||
| AK165439.1 | - | 10 - 11 | 1224 | 71 | ||
| BC013528.1 | - | 10 - 11 | 1217 | 4238222 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a1
Genomic Map