Assay Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011539.3 | NP_035669.3 | 5 - 6 | 616 | 72 | |
| GenBank mRNA | AK048544.1 | - | 5 - 6 | 598 | 72 | |
| AK085414.1 | - | 2 - 3 | 367 | 72 | ||
| AK139479.1 | - | 5 - 6 | 579 | 72 | ||
| BC029014.1 | - | 5 - 6 | 618 | 5007629 | 72 | |
| L18868.1 | - | 5 - 6 | 487 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbxas1
Genomic Map