Assay Details
| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122948.2 | NP_001116420.1 | 2 - 3 | 855 | 116 | |
| NM_001301674.1 | NP_001288603.1 | 3 - 4 | 834 | 116 | ||
| NM_011547.4 | NP_035677.2 | 2 - 3 | 590 | 116 | ||
| GenBank mRNA | AB221578.1 | - | 2 - 3 | 481 | 116 | |
| AK053704.1 | - | 2 - 3 | 840 | 116 | ||
| AK145772.1 | - | 2 - 3 | 545 | 116 | ||
| BC007471.1 | - | 2 - 3 | 565 | 3497082 | 116 | |
| BC018226.1 | - | 3 - 4 | 833 | 3983850 | 116 | |
| U17285.1 | - | 2 - 3 | 582 | 116 | ||
| U17287.1 | - | 2 - 3 | 819 | 116 | ||
| U17288.1 | - | 1 - 2 | 604 | 116 | ||
| X74216.1 | - | 2 - 3 | 551 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfap2a
Genomic Map