Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 6 - 7 | 1522 | 99 | |
| GenBank mRNA | AF230391.1 | - | 6 - 7 | 1123 | 99 | |
| AF230392.1 | - | 6 - 7 | 1123 | 99 | ||
| AK141357.1 | - | 6 - 7 | 1266 | 99 | ||
| AK149995.1 | - | 3 - 4 | 347 | 99 | ||
| BC020452.1 | - | 6 - 7 | 953 | 99 | ||
| BC023335.1 | - | 6 - 7 | 1231 | 99 | ||
| BC058391.1 | - | 6 - 7 | 968 | 6817165 | 99 | |
| BC089337.1 | - | 6 - 7 | 1522 | 6811294 | 99 | |
| U67303.1 | - | 6 - 7 | 1513 | 99 | ||
| X99644.1 | - | 6 - 7 | 1123 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map