Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 12 - 13 | 2222 | 62 | |
| GenBank mRNA | AF230391.1 | - | 12 - 13 | 1823 | 62 | |
| AK141357.1 | - | 12 - 13 | 1966 | 62 | ||
| AK149995.1 | - | 9 - 10 | 1047 | 62 | ||
| BC020452.1 | - | 12 - 13 | 1653 | 62 | ||
| BC023335.1 | - | 12 - 13 | 1931 | 62 | ||
| BC058391.1 | - | 12 - 13 | 1668 | 6817165 | 62 | |
| BC089337.1 | - | 12 - 13 | 2222 | 6811294 | 62 | |
| U67303.1 | - | 12 - 13 | 2213 | 62 | ||
| X99644.1 | - | 12 - 13 | 1823 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map