Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 14 - 15 | 2678 | 68 | |
| GenBank mRNA | AF230391.1 | - | 14 - 15 | 2279 | 68 | |
| AK089084.1 | - | 2 - 3 | 330 | 68 | ||
| AK141357.1 | - | 14 - 15 | 2422 | 68 | ||
| AK149995.1 | - | 11 - 12 | 1503 | 68 | ||
| BC020452.1 | - | 14 - 15 | 2109 | 68 | ||
| BC023335.1 | - | 14 - 15 | 2387 | 68 | ||
| BC058391.1 | - | 14 - 15 | 2124 | 6817165 | 68 | |
| BC089337.1 | - | 14 - 15 | 2678 | 6811294 | 68 | |
| U67303.1 | - | 14 - 15 | 2669 | 68 | ||
| X99644.1 | - | 14 - 15 | 2279 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map