Assay Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011588.3 | NP_035718.2 | 15 - 16 | 2760 | 65 | |
| GenBank mRNA | AF230391.1 | - | 15 - 16 | 2361 | 65 | |
| AK089084.1 | - | 3 - 4 | 412 | 65 | ||
| AK141357.1 | - | 15 - 16 | 2504 | 65 | ||
| AK149995.1 | - | 12 - 13 | 1585 | 65 | ||
| AK183204.1 | - | 1 - 2 | 90 | 65 | ||
| BC020452.1 | - | 15 - 16 | 2191 | 65 | ||
| BC023335.1 | - | 15 - 16 | 2469 | 65 | ||
| BC058391.1 | - | 15 - 16 | 2206 | 6817165 | 65 | |
| BC089337.1 | - | 15 - 16 | 2760 | 6811294 | 65 | |
| U67303.1 | - | 15 - 16 | 2751 | 65 | ||
| X99644.1 | - | 15 - 16 | 2361 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim28
Genomic Map