Assay Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198985.1 | NP_001185914.1 | 11 - 12 | 1806 | 62 | |
| NM_011597.4 | NP_035727.2 | 11 - 12 | 1796 | 62 | ||
| GenBank mRNA | AF113005.1 | - | 11 - 12 | 1654 | 62 | |
| BC034677.1 | - | 11 - 12 | 1714 | 3155078 | 62 | |
| BC039924.1 | - | 11 - 12 | 1725 | 3155079 | 62 | |
| BC042695.1 | - | 11 - 12 | 1698 | 62 | ||
| CN527767.1 | - | 3 - 4 | 339 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tjp2
Genomic Map