Assay Details
| Gene Symbol: Tle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285529.1 | NP_001272458.1 | 2 - 3 | 1010 | 82 | |
| NM_001285530.1 | NP_001272459.1 | 2 - 3 | 1010 | 82 | ||
| NM_001285531.1 | NP_001272460.1 | 2 - 3 | 1010 | 82 | ||
| NM_001285532.1 | NP_001272461.1 | 2 - 3 | 1010 | 82 | ||
| NM_011599.5 | NP_035729.3 | 2 - 3 | 1010 | 82 | ||
| GenBank mRNA | AK044642.1 | - | 2 - 3 | 1012 | 82 | |
| AK046402.1 | - | 2 - 3 | 978 | 82 | ||
| AK048515.1 | - | 2 - 3 | 996 | 82 | ||
| AK076750.1 | - | 2 - 3 | 392 | 82 | ||
| AK082499.1 | - | 2 - 3 | 976 | 82 | ||
| AY155196.1 | - | 2 - 3 | 979 | 82 | ||
| AY155197.1 | - | 2 - 3 | 168 | 82 | ||
| AY155198.1 | - | 2 - 3 | 169 | 82 | ||
| AY155199.1 | - | 2 - 3 | 169 | 82 | ||
| BC057573.1 | - | 2 - 3 | 840 | 6401650 | 82 | |
| U61362.1 | - | 2 - 3 | 176 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tle1
Genomic Map