Assay Details
| Gene Symbol: Tom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011622.3 | NP_035752.1 | 10 - 11 | 1121 | 67 | |
| GenBank mRNA | AJ006972.1 | - | 10 - 11 | 1098 | 67 | |
| AK028398.1 | - | 10 - 11 | 1062 | 67 | ||
| AK051245.1 | - | 10 - 11 | 1103 | 67 | ||
| AK160403.1 | - | 10 - 11 | 1067 | 67 | ||
| AK182481.1 | - | 1 - 2 | 45 | 67 | ||
| BC021633.1 | - | 10 - 11 | 1084 | 4208226 | 67 | |
| BC093520.1 | - | 10 - 11 | 1049 | 2866013 | 67 | |
| BC157905.1 | - | 10 - 11 | 1082 | 9088106 | 67 | |
| BC158042.1 | - | 10 - 11 | 1082 | 9007343 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tom1
Genomic Map