Assay Details
| Gene Symbol: Trap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011635.1 | NP_035765.1 | 1 - 2 | 729 | 103 | |
| GenBank mRNA | AK146737.1 | - | 1 - 2 | 544 | 103 | |
| BC051644.1 | - | 1 - 2 | 522 | 30022193 | 103 | |
| M36387.1 | - | 1 - 2 | 729 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32693896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32693895 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | NM_011635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | AK146737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | BC051644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | M36387.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | NM_011635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | AK146737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | BC051644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P537571 | M36387.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10883 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | NM_011635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | AK146737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | BC051644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | M36387.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | NM_011635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | AK146737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | BC051644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P537571 | M36387.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32693734 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435731 | - | - |
| Whole Mouse Genome G4122A | A_52_P537571 | NM_011635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | AK146737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | BC051644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | M36387.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | NM_011635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | AK146737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | BC051644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537571 | M36387.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15137103 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15137095 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15137097 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15137099 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15137101 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14756647 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32693895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32693896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32693764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14979222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15137102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14956586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15086055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15060260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14878106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14756647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14781157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14979223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14956589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14567301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14842119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32693761 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601990 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460226_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601988 | - | - |
| Mouse Genome 430 2.0 Array | 1460226_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460226_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22037_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22037_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00008600.mm.1 | NM_011635.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008600.mm.1 | TC0X00001368.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008601.mm.1 | NM_011635.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008601.mm.1 | TC0X00001368.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008602.mm.1 | NM_011635.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008602.mm.1 | TC0X00001368.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460226_at | - | - |
| Mouse Expression Set 430 | 1460226_PM_at | - | - |
| Murine Genome U74v2 Set | 102764_at | - | - |
| Murine Genome U74v2 Set | 102764_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460226_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460226_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460226_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460226_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 741395 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4810408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2463395 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2463395 | - | - |
| mouse-6 v1.1 (Illumina) | scl54594.3.1_69-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54594.3.1_69-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2463395 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2463395 | - | - |

Trap1a
Genomic Map