Assay Details
| Gene Symbol: Trp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127259.1 | NP_001120731.1 | 7 - 8 | 1517 | 93 | |
| NM_001127260.1 | NP_001120732.1 | 7 - 8 | 1517 | 93 | ||
| NM_001127261.1 | NP_001120733.1 | 7 - 8 | 1517 | 93 | ||
| NM_001127262.1 | NP_001120734.1 | 5 - 6 | 854 | 93 | ||
| NM_001127263.1 | NP_001120735.1 | 5 - 6 | 854 | 93 | ||
| NM_001127264.1 | NP_001120736.1 | 5 - 6 | 854 | 93 | ||
| NM_001127265.1 | NP_001120737.1 | 5 - 6 | 854 | 93 | ||
| NM_011641.2 | NP_035771.1 | 5 - 6 | 854 | 93 | ||
| GenBank mRNA | AB010152.1 | - | 5 - 6 | 828 | 93 | |
| AF075434.1 | - | 7 - 8 | 992 | 93 | ||
| AF075435.1 | - | 7 - 8 | 992 | 93 | ||
| AF075436.1 | - | 7 - 8 | 992 | 93 | ||
| AF075437.1 | - | 5 - 6 | 710 | 93 | ||
| AF075438.1 | - | 5 - 6 | 710 | 93 | ||
| AF075439.1 | - | 5 - 6 | 710 | 93 | ||
| AK048623.1 | - | 5 - 6 | 854 | 93 | ||
| AK137253.1 | - | 5 - 6 | 855 | 93 | ||
| BC090649.1 | - | 5 - 6 | 836 | 30645427 | 93 | |
| BC092537.1 | - | 5 - 6 | 854 | 30504012 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp63
Genomic Map