Assay Details
| Gene Symbol: Trp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127259.1 | NP_001120731.1 | 12 - 13 | 2175 | 109 | |
| NM_001127264.1 | NP_001120736.1 | 10 - 11 | 1500 | 109 | ||
| NM_011641.2 | NP_035771.1 | 10 - 11 | 1512 | 109 | ||
| GenBank mRNA | AB010152.1 | - | 10 - 11 | 1486 | 109 | |
| AF075436.1 | - | 12 - 13 | 1650 | 109 | ||
| AF075439.1 | - | 10 - 11 | 1368 | 109 | ||
| AK137253.1 | - | 10 - 11 | 1501 | 109 | ||
| BC090649.1 | - | 10 - 11 | 1482 | 30645427 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trp63
Genomic Map