Assay Details
| Gene Symbol: Tubb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011655.5 | NP_035785.1 | 1 - 2 | 305 | 94 | |
| GenBank mRNA | AK011263.1 | - | 1 - 2 | 233 | 94 | |
| AK011765.1 | - | 1 - 2 | 237 | 94 | ||
| AK012350.1 | - | 1 - 2 | 237 | 94 | ||
| AK017806.1 | - | 1 - 2 | 237 | 94 | ||
| AK051164.1 | - | 1 - 2 | 237 | 94 | ||
| AK051393.1 | - | 1 - 2 | 239 | 94 | ||
| AK083327.1 | - | 1 - 2 | 237 | 94 | ||
| AK146567.1 | - | 1 - 2 | 237 | 94 | ||
| AK148978.1 | - | 1 - 2 | 232 | 94 | ||
| AK151215.1 | - | 1 - 2 | 237 | 94 | ||
| AK151670.1 | - | 1 - 2 | 237 | 94 | ||
| AK160225.1 | - | 1 - 2 | 237 | 94 | ||
| AK161575.1 | - | 1 - 2 | 237 | 94 | ||
| AK165249.1 | - | 1 - 2 | 235 | 94 | ||
| AK167691.1 | - | 1 - 2 | 237 | 94 | ||
| AK167779.1 | - | 1 - 2 | 237 | 94 | ||
| AK167808.1 | - | 1 - 2 | 232 | 94 | ||
| AK167926.1 | - | 1 - 2 | 237 | 94 | ||
| AK168271.1 | - | 1 - 2 | 232 | 94 | ||
| AK169123.1 | - | 1 - 2 | 232 | 94 | ||
| AK169210.1 | - | 1 - 2 | 236 | 94 | ||
| AK169295.1 | - | 1 - 2 | 237 | 94 | ||
| AK169606.1 | - | 1 - 2 | 238 | 94 | ||
| BC003825.1 | - | 1 - 2 | 211 | 3601177 | 94 | |
| X04663.1 | - | 1 - 2 | 94 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tubb5
Genomic Map