Assay Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011672.4 | NP_035802.3 | 2 - 3 | 255 | 78 | |
| GenBank mRNA | AK010461.1 | - | 2 - 3 | 224 | 78 | |
| AK088823.1 | - | 2 - 3 | 224 | 78 | ||
| AK147138.1 | - | 2 - 3 | 238 | 78 | ||
| AK169062.1 | - | 2 - 3 | 237 | 78 | ||
| BC006630.1 | - | 2 - 3 | 212 | 3708320 | 78 | |
| CJ085208.1 | - | 2 - 3 | 255 | 78 | ||
| U64445.1 | - | 2 - 3 | 240 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufd1l
Genomic Map