Assay Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011672.4 | NP_035802.3 | 4 - 5 | 412 | 78 | |
| NR_028403.1 | 4 - 5 | 386 | 78 | |||
| GenBank mRNA | AK010461.1 | - | 4 - 5 | 381 | 78 | |
| AK147138.1 | - | 4 - 5 | 395 | 78 | ||
| AK169062.1 | - | 4 - 5 | 394 | 78 | ||
| BC006630.1 | - | 4 - 5 | 369 | 3708320 | 78 | |
| U64445.1 | - | 4 - 5 | 397 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufd1l
Genomic Map