Assay Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011672.4 | NP_035802.3 | 7 - 8 | 681 | 73 | |
| NR_028403.1 | 7 - 8 | 655 | 73 | |||
| GenBank mRNA | AK010461.1 | - | 7 - 8 | 650 | 73 | |
| AK012543.1 | - | 5 - 6 | 625 | 73 | ||
| AK088823.1 | - | 7 - 8 | 650 | 73 | ||
| AK147138.1 | - | 7 - 8 | 664 | 73 | ||
| AK169062.1 | - | 7 - 8 | 663 | 73 | ||
| BC006630.1 | - | 7 - 8 | 638 | 3708320 | 73 | |
| U64445.1 | - | 7 - 8 | 666 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufd1l
Genomic Map