Assay Details
| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011673.3 | NP_035803.1 | 3 - 4 | 437 | 99 | |
| GenBank mRNA | AK138848.1 | - | 3 - 4 | 453 | 99 | |
| BC050828.1 | - | 3 - 4 | 437 | 6512920 | 99 | |
| D89866.1 | - | 3 - 4 | 528 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ugcg
Genomic Map