Assay Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011678.2 | NP_035808.2 | 2 - 3 | 311 | 67 | |
| GenBank mRNA | AK138838.1 | - | 2 - 3 | 246 | 67 | |
| AK143582.1 | - | 2 - 3 | 246 | 67 | ||
| AK149964.1 | - | 2 - 3 | 269 | 67 | ||
| AK169933.1 | - | 2 - 3 | 281 | 67 | ||
| AK171271.1 | - | 2 - 3 | 281 | 67 | ||
| AK171633.1 | - | 2 - 3 | 264 | 67 | ||
| AK214431.1 | - | 1 - 2 | 107 | 67 | ||
| AK220512.1 | - | 2 - 3 | 268 | 67 | ||
| BC066180.1 | - | 2 - 3 | 221 | 67 | ||
| BC066865.1 | - | 2 - 3 | 221 | 67 | ||
| L00681.1 | - | 2 - 3 | 274 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp4
Genomic Map