Assay Details
| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293758.1 | NP_001280687.1 | 2 - 3 | 302 | 78 | |
| NM_011722.4 | NP_035852.1 | 2 - 3 | 356 | 78 | ||
| GenBank mRNA | AF124788.1 | - | 2 - 3 | 87 | 78 | |
| AF190796.1 | - | 2 - 3 | 97 | 78 | ||
| AK003618.1 | - | 2 - 3 | 96 | 78 | ||
| AK088312.1 | - | 2 - 3 | 103 | 78 | ||
| BC029249.1 | - | 2 - 3 | 97 | 4486579 | 78 | |
| BY079663.1 | - | 2 - 3 | 156 | 78 | ||
| CA573939.1 | - | 2 - 3 | 356 | 78 | ||
| DV042569.1 | - | 2 - 3 | 311 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dctn6
Genomic Map