Assay Details
| Gene Symbol: Gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011818.3 | NP_035948.3 | 7 - 8 | 1053 | 92 | |
| GenBank mRNA | AF163665.1 | - | 7 - 8 | 997 | 92 | |
| AF186095.1 | - | 7 - 8 | 889 | 92 | ||
| AF282322.1 | - | 7 - 8 | 923 | 92 | ||
| AK004716.1 | - | 7 - 8 | 1029 | 92 | ||
| AK146964.1 | - | 7 - 8 | 1022 | 92 | ||
| AK155238.1 | - | 7 - 8 | 1036 | 92 | ||
| BC054767.1 | - | 7 - 8 | 1012 | 6414943 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmcl1
Genomic Map