Assay Details
| Gene Symbol: Lynx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011838.4 | NP_035968.1 | 2 - 3 | 278 | 85 | |
| GenBank mRNA | AF141377.1 | - | 2 - 3 | 252 | 85 | |
| AK013827.1 | - | 2 - 3 | 275 | 85 | ||
| AK161907.1 | - | 2 - 3 | 308 | 85 | ||
| AK162920.1 | - | 2 - 3 | 301 | 85 | ||
| AK165295.1 | - | 2 - 3 | 282 | 85 | ||
| AK165314.1 | - | 2 - 3 | 282 | 85 | ||
| BC037541.1 | - | 2 - 3 | 325 | 5325123 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lynx1
Genomic Map