Assay Details
| Gene Symbol: Tekt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011902.2 | NP_036032.2 | 1 - 2 | 71 | 72 | |
| GenBank mRNA | AB027138.1 | - | 1 - 2 | 71 | 72 | |
| BC008140.1 | - | 1 - 2 | 87 | 3582971 | 72 | |
| BQ712964.1 | - | 1 - 2 | 85 | 72 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB027138.1,BC008140.1,NM_011902.2 transcripts.
Set Membership: |
|

Tekt2
Genomic Map