Assay Details
| Gene Symbol: Decr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011933.2 | NP_036063.1 | 4 - 5 | 460 | 74 | |
| GenBank mRNA | AF155575.1 | - | 4 - 5 | 460 | 74 | |
| AK137212.1 | - | 4 - 5 | 391 | 74 | ||
| AK149270.1 | - | 4 - 5 | 585 | 74 | ||
| AK162961.1 | - | 4 - 5 | 596 | 74 | ||
| BC021865.1 | - | 4 - 5 | 432 | 5123899 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129002 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133081 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108243 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08744504 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133081 | - | - |
| Whole Mouse Genome G4122A | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | BC021865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AK162961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AF155575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AK137212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P443293 | AK149270.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109113 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138800 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173432 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173439 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173433 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173436 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13173438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13411640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13579138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13553668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13382154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13265973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13106700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13553673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13307958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13106698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13382159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13440988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13173438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13106704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13106701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108168 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449235 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423495_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449225 | - | - |
| Mouse Genome 430 2.0 Array | 1423495_at | - | - |
| Mouse Genome 430 2.0 Array | 1423495_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26378_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26378_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700015672.mm.1 | NM_011933.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015672.mm.1 | NM_011933.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015672.mm.1 | TC1700001688.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423495_at | - | - |
| Mouse Expression Set 430 | 1423495_PM_at | - | - |
| Murine Genome U74v2 Set | 131478_at | - | - |
| Murine Genome U74v2 Set | 131478_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423495_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423495_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423495_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423495_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 716213 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1980500 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2656386 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2656386 | - | - |
| mouse-6 v1.1 (Illumina) | scl50155.9_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001610.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980500 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50155.9_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001610.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520280 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2705849 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2705848 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2705848 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2705849 | - | - |

Decr2
Genomic Map