Assay Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011993.4 | NP_036123.3 | 7 - 8 | 816 | 73 | |
| GenBank mRNA | AB006715.1 | - | 7 - 8 | 745 | 73 | |
| AK090197.1 | - | 6 - 7 | 1343 | 73 | ||
| AK145638.1 | - | 6 - 7 | 919 | 73 | ||
| AK165694.1 | - | 7 - 8 | 817 | 73 | ||
| BC046333.1 | - | 6 - 7 | 2369 | 73 | ||
| BC090674.1 | - | 788 | 72 | |||
| BC138573.1 | - | 7 - 8 | 734 | 8861593 | 73 | |
| BC138574.1 | - | 7 - 8 | 734 | 8861594 | 73 | |
| Y09079.1 | - | 7 - 8 | 795 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl4
Genomic Map