Assay Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011993.4 | NP_036123.3 | 9 - 10 | 1085 | 80 | |
| GenBank mRNA | AB006715.1 | - | 9 - 10 | 1014 | 80 | |
| AK090197.1 | - | 8 - 9 | 1613 | 80 | ||
| AK145638.1 | - | 8 - 9 | 1188 | 80 | ||
| AK165694.1 | - | 9 - 10 | 1086 | 80 | ||
| BC046333.1 | - | 8 - 9 | 2638 | 80 | ||
| BC090674.1 | - | 1057 | 80 | |||
| BC138573.1 | - | 9 - 10 | 1003 | 8861593 | 80 | |
| BC138574.1 | - | 9 - 10 | 1003 | 8861594 | 80 | |
| Y09079.1 | - | 9 - 10 | 1064 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl4
Genomic Map