Assay Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013456.2 | NP_038484.1 | 12 - 13 | 1477 | 87 | |
| GenBank mRNA | AF093775.1 | - | 12 - 13 | 1452 | 87 | |
| BC111890.1 | - | 12 - 13 | 1443 | 40060598 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actn3
Genomic Map